missing translation for 'onlineSavingsMsg'
Learn More

Sucrose, 99% max., Molecular biology reagent grade, MP Biomedicals™

Catalog No. ICN19401891
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19401891 1 kg
ICN19401894 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19401891 Supplier MP Biomedicals Inc Supplier No. 0219401891

Chemical Identifiers

CAS 57-50-1
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006626
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
Synonym sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
PubChem CID 5988
ChEBI CHEBI:17992
IUPAC Name (2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

Specifications

Color White
Density 1.587g/cm3 at 25°C
Assay Percent Range >99%
Quantity 1 kg
Formula Weight 342.3
Grade Molecular Biology Reagent
Physical Form Crystalline Powder
Chemical Name or Material Sucrose
EINECSNumber 200-334-9
Recommended Storage Store at room temperature.