missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals Inc Sucrose, 99% max., Molecular biology reagent grade, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0219401894

 View more versions of this product

Catalog No. ICN19401894



Description

Description

Specifications

Specifications

Sucrose
White
>99%
5 kg
CZMRCDWAGMRECN-UGDNZRGBSA-N
(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
5988
342.3
Crystalline Powder
57-50-1
1.587g/cm3 at 25°C
C12H22O11
sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O
342.297
CHEBI:17992
Molecular Biology Reagent

Chemical Identifiers

57-50-1
342.297
sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
CHEBI:17992
C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O
C12H22O11
CZMRCDWAGMRECN-UGDNZRGBSA-N
5988
(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
Safety and Handling

Safety and Handling

EINECSNumber : 200-334-9

Recommended Storage : Store at room temperature.

SDS
Documents

Documents

Promotions

Promotions