accessibility menu, dialog, popup
UserName
Fisher Scientific Logo
EN
chevron-down
Select Language
English
Français
Search
Order Status
Quick Order
Support
Offers
chevron-down
Featured
Spotlight
Offers
Flex Dollars
New Lab Start-up
View All
Sign In
chevron-down
chevron-down
Sign In
Don't have an account?
Create an Account
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Biochemical Reagents
Carbohydrates
Glycosyl compounds
Sucrose, ≥99%, Cell Culture Reagent, MP Biomedicals™
Structure Search
Print
Sucrose, ≥99%, Cell Culture Reagent, MP Biomedicals™
Catalog No.
ICN19474701
Change view
Chip view icon
List view icon
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
Quantity
ICN19474701
1 kg
ICN19474705
5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No.
ICN19474701
Supplier
MP Biomedicals Inc
Supplier No.
0219474701
Sign In
or
Register
to check your price and availability.
$129.01
/
Each
Add to Cart
Add to Cart
Description
Specifications
Product Suggestions
Videos
Documents
Certificates
Safety and Handling
Safety Services
Promotions
Specifications
Chemical Identifiers
CAS
57-50-1
Molecular Formula
C12H22O11
Molecular Weight (g/mol)
342.30
MDL Number
MFCD00006626
InChI Key
CZMRCDWAGMRECN-PWPRYFECNA-N
Synonym
sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
PubChem CID
5988
ChEBI
CHEBI:17992
IUPAC Name
(2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES
OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
Specifications
Quantity
1 kg
Product Suggestions
Documents