missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals Inc Sucrose, 99.97%, MP Biomedicals™
Supplier: MP Biomedicals Inc 0219027101
Specifications
Sucrose | |
White | |
99.7% | |
1 kg | |
CZMRCDWAGMRECN-UGDNZRGBSA-N | |
(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol | |
5988 | |
342.3 |
57-50-1 | |
1.587g/cm3 at 25°C | |
C12H22O11 | |
sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand | |
C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O | |
342.297 | |
CHEBI:17992 | |
Crystalline Powder |
Chemical Identifiers
57-50-1 | |
342.297 | |
sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand | |
CHEBI:17992 | |
C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O |
C12H22O11 | |
CZMRCDWAGMRECN-UGDNZRGBSA-N | |
5988 | |
(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
Safety and Handling
EINECSNumber : 200-334-9
Recommended Storage : Store at room temperature.