missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Sucralose | |
| 130°C | |
| C12H19Cl3O8 | |
| 25 g | |
| BAQAVOSOZGMPRM-QBMZZYIRSA-N | |
| (2R,3R,4R,5R,6R)-2-{[(2R,3S,4S,5S)-2,5-bis(chloromethyl)-3,4-dihydroxyoxolan-2-yl]oxy}-5-chloro-6-(hydroxymethyl)oxane-3,4-diol | |
| 71485 | |
| 397.63 | |
| Crystalline Powder |
| 56038-13-2 | |
| White | |
| MFCD03648615 | |
| sucralose, trichlorosucrose, splenda, 1',4,6'-trichlorogalactosucrose, unii-96k6uq3zd4, 1,6-dichloro-1,6-dideoxy-beta-d-fructofuranosyl 4-chloro-4-deoxy-alpha-d-galactopyranoside, 1,6-dichloro-1,6-dideoxy-beta-d-fructofuranosyl-4-chloro-4-deoxy-alpha-d-galactopyranoside, trichlorogalactosucrose, trichlorogalacto-sucrose, aspasvit | |
| OC[C@H]1O[C@H](O[C@]2(CCl)O[C@H](CCl)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@H]1Cl | |
| 397.63 | |
| CHEBI:32159 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 56038-13-2 | |
| 397.63 | |
| BAQAVOSOZGMPRM-QBMZZYIRSA-N | |
| 71485 | |
| (2R,3R,4R,5R,6R)-2-{[(2R,3S,4S,5S)-2,5-bis(chloromethyl)-3,4-dihydroxyoxolan-2-yl]oxy}-5-chloro-6-(hydroxymethyl)oxane-3,4-diol |
| C12H19Cl3O8 | |
| MFCD03648615 | |
| sucralose, trichlorosucrose, splenda, 1',4,6'-trichlorogalactosucrose, unii-96k6uq3zd4, 1,6-dichloro-1,6-dideoxy-beta-d-fructofuranosyl 4-chloro-4-deoxy-alpha-d-galactopyranoside, 1,6-dichloro-1,6-dideoxy-beta-d-fructofuranosyl-4-chloro-4-deoxy-alpha-d-galactopyranoside, trichlorogalactosucrose, trichlorogalacto-sucrose, aspasvit | |
| CHEBI:32159 | |
| OC[C@H]1O[C@H](O[C@]2(CCl)O[C@H](CCl)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@H]1Cl |
Safety and Handling
RTECSNumber : LW5440140
TSCA : No