Learn More
Thermo Scientific Chemicals Sucralose, 98%
CAS: 56038-13-2 | C12H19Cl3O8 | 397.63 g/mol
Supplier: Thermo Scientific Chemicals J6673630
Description
Sucralose is a non-caloric sugar used as a sweetener that activates T1R2/T1R3 sweet taste receptors and modulates glucagon-like peptide-1, glucose-dependent insulinotrophic peptide, and incretin secretion.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSucralose is a non-caloric sugar used as a sweetener that activates T1R2/T1R3 sweet taste receptors and modulates glucagon-like peptide-1, glucose-dependent insulinotrophic peptide, and incretin secretion.
Solubility
Soluble in Water.
Notes
Research use only: Not intended for food, cosmetic, or diagnostic and therapeutic use in animals nor humans.
Specifications
| Sucralose | |
| 130°C (decomposition) | |
| C12H19Cl3O8 | |
| 250 g | |
| 14,8880 | |
| Soluble in Water. | |
| OC[C@H]1O[C@H](O[C@]2(CCl)O[C@H](CCl)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@H]1Cl | |
| 397.63 | |
| CHEBI:32159 | |
| 98% |
| 56038-13-2 | |
| White | |
| MFCD03648615 | |
| 3654410 | |
| sucralose, trichlorosucrose, splenda, 1',4,6'-trichlorogalactosucrose, unii-96k6uq3zd4, 1,6-dichloro-1,6-dideoxy-beta-d-fructofuranosyl 4-chloro-4-deoxy-alpha-d-galactopyranoside, 1,6-dichloro-1,6-dideoxy-beta-d-fructofuranosyl-4-chloro-4-deoxy-alpha-d-galactopyranoside, trichlorogalactosucrose, trichlorogalacto-sucrose, aspasvit | |
| BAQAVOSOZGMPRM-QBMZZYIRSA-N | |
| (2R,3R,4R,5R,6R)-2-[(2R,3S,4S,5S)-2,5-bis(chloromethyl)-3,4-dihydroxyoxolan-2-yl]oxy-5-chloro-6-(hydroxymethyl)oxane-3,4-diol | |
| 71485 | |
| 397.63 | |
| Powder |
Chemical Identifiers
| 56038-13-2 | |
| 397.63 | |
| BAQAVOSOZGMPRM-QBMZZYIRSA-N | |
| 71485 | |
| (2R,3R,4R,5R,6R)-2-[(2R,3S,4S,5S)-2,5-bis(chloromethyl)-3,4-dihydroxyoxolan-2-yl]oxy-5-chloro-6-(hydroxymethyl)oxane-3,4-diol |
| C12H19Cl3O8 | |
| MFCD03648615 | |
| sucralose, trichlorosucrose, splenda, 1',4,6'-trichlorogalactosucrose, unii-96k6uq3zd4, 1,6-dichloro-1,6-dideoxy-beta-d-fructofuranosyl 4-chloro-4-deoxy-alpha-d-galactopyranoside, 1,6-dichloro-1,6-dideoxy-beta-d-fructofuranosyl-4-chloro-4-deoxy-alpha-d-galactopyranoside, trichlorogalactosucrose, trichlorogalacto-sucrose, aspasvit | |
| CHEBI:32159 | |
| OC[C@H]1O[C@H](O[C@]2(CCl)O[C@H](CCl)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@H]1Cl |
Safety and Handling
EINECSNumber : 259-952-2
RTECSNumber : LW5440140
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only