missing translation for 'onlineSavingsMsg'
Learn More

Succinylcholine Chloride Dihydrate 98.0+%, TCI America™

Catalog No. S01495G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S01495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S01495G Supplier TCI America Supplier No. S01495G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6101-15-1
Molecular Formula C14H34Cl2N2O6
Molecular Weight (g/mol) 397.33
MDL Number MFCD00150564
InChI Key FFSBEIRFVXGRPR-UHFFFAOYSA-L
Synonym Suxamethonium Chloride
PubChem CID 656867
ChEBI CHEBI:61225
IUPAC Name trimethyl[2-({4-oxo-4-[2-(trimethylazaniumyl)ethoxy]butanoyl}oxy)ethyl]azanium dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].C[N+](C)(C)CCOC(=O)CCC(=O)OCC[N+](C)(C)C

Specifications

Melting Point 164°C
Color White
Quantity 5 g
UN Number 2811
Formula Weight 361.30
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Succinylcholine Chloride Dihydrate
RTECSNumber GA2360000
TSCA No