missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Succinyl Chloride 95.0+%, TCI America™
$48.13 - $444.72
Chemical Identifiers
| CAS | 543-20-4 |
|---|---|
| Molecular Formula | C4H4Cl2O2 |
| Molecular Weight (g/mol) | 154.974 |
| MDL Number | MFCD00000749 |
| InChI Key | IRXBNHGNHKNOJI-UHFFFAOYSA-N |
| Synonym | succinyl chloride, succinyl dichloride, succinic acid dichloride, succinoyl chloride, succinoyl dichloride, succinic chloride, 1,2-bis chlorocarbonyl ethane, butanedioyl chloride, unii-gdn09v9867, 1,2-ethanediylbis chloroformate |
| PubChem CID | 10970 |
| IUPAC Name | butanedioyl dichloride |
| SMILES | C(CC(=O)Cl)C(=O)Cl |
Chemical Identifiers
| 543-20-4 | |
| 154.974 | |
| IRXBNHGNHKNOJI-UHFFFAOYSA-N | |
| 10970 | |
| C(CC(=O)Cl)C(=O)Cl |
| C4H4Cl2O2 | |
| MFCD00000749 | |
| succinyl chloride, succinyl dichloride, succinic acid dichloride, succinoyl chloride, succinoyl dichloride, succinic chloride, 1,2-bis chlorocarbonyl ethane, butanedioyl chloride, unii-gdn09v9867, 1,2-ethanediylbis chloroformate | |
| butanedioyl dichloride |
Specifications
| 543-20-4 | |
| 192°C | |
| MFCD00000749 | |
| 3265 | |
| IRXBNHGNHKNOJI-UHFFFAOYSA-N | |
| butanedioyl dichloride | |
| 10970 | |
| ≥95.0% (T) | |
| Succinyl Chloride |
| Yellow | |
| C4H4Cl2O2 | |
| 25 g | |
| succinyl chloride, succinyl dichloride, succinic acid dichloride, succinoyl chloride, succinoyl dichloride, succinic chloride, 1,2-bis chlorocarbonyl ethane, butanedioyl chloride, unii-gdn09v9867, 1,2-ethanediylbis chloroformate | |
| C(CC(=O)Cl)C(=O)Cl | |
| 154.974 | |
| 154.97 | |
| Liquid |
Safety and Handling
RTECSNumber : WN4900000
TSCA : Yes