missing translation for 'onlineSavingsMsg'
Learn More

Succinyl Chloride, ca. 95%

CAS: 543-20-4 | C4H4Cl2O2 | 154.98 g/mol

$83.62 - $83.62

Chemical Identifiers

CAS 543-20-4
Molecular Formula C4H4Cl2O2
Molecular Weight (g/mol) 154.98
MDL Number MFCD00000749
InChI Key IRXBNHGNHKNOJI-UHFFFAOYSA-N
Synonym succinyl chloride, succinyl dichloride, succinic acid dichloride, succinoyl chloride, succinoyl dichloride, succinic chloride, 1,2-bis chlorocarbonyl ethane, butanedioyl chloride, unii-gdn09v9867, 1,2-ethanediylbis chloroformate
PubChem CID 10970
IUPAC Name butanedioyl dichloride
SMILES C(CC(=O)Cl)C(=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC132840250
SDS View Documents
Thermo Scientific Chemicals
132840250
25 g Glass bottle
Each for $83.62
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

543-20-4
154.98
IRXBNHGNHKNOJI-UHFFFAOYSA-N
10970
C(CC(=O)Cl)C(=O)Cl
C4H4Cl2O2
MFCD00000749
succinyl chloride, succinyl dichloride, succinic acid dichloride, succinoyl chloride, succinoyl dichloride, succinic chloride, 1,2-bis chlorocarbonyl ethane, butanedioyl chloride, unii-gdn09v9867, 1,2-ethanediylbis chloroformate
butanedioyl dichloride

Specifications

543-20-4
Colorless to Brown
190°C
Authentic
Glass bottle
1.4700 to 1.472
MFCD00000749
02, IV, 1921
15, 9004
Solubility in water: decomposes
C(CC(=O)Cl)C(=O)Cl
154.98
154.98
Liquid
16°C to 17°C
1.4070g/mL
76°C
ca. 95%
C4H4Cl2O2
ClCOCH2CH2COCl
25 g
1.407
succinyl chloride, succinyl dichloride, succinic acid dichloride, succinoyl chloride, succinoyl dichloride, succinic chloride, 1,2-bis chlorocarbonyl ethane, butanedioyl chloride, unii-gdn09v9867, 1,2-ethanediylbis chloroformate
IRXBNHGNHKNOJI-UHFFFAOYSA-N
butanedioyl dichloride
10970
ca. 95%
Succinyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with

GHS Signal Word: Danger

EINECSNumber : 208-838-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only