missing translation for 'onlineSavingsMsg'
Learn More

Succinyl chloride, 96%

Catalog No. AAA1838918
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1838918 50 g
AAA1838930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1838918 Supplier Thermo Scientific Chemicals Supplier No. A1838918

CAS: 543-20-4 | C4H4Cl2O2 | 154.974 g/mol

It is employed in the synthesis of biseugenyl succinate and bis(4-maleimidephenyl) succinate. It is also used as used as an important raw material and intermediate in pharmaceuticals, organic synthesis, agrochemicals and dye stuffs. It is also applied as hydrolytic reagent for the determination of water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis of biseugenyl succinate and bis(4-maleimidephenyl) succinate. It is also used as used as an important raw material and intermediate in pharmaceuticals, organic synthesis, agrochemicals and dye stuffs. It is also applied as hydrolytic reagent for the determination of water.

Solubility
Reacts violently with water.

Notes
Moisture Sensitive. Store away from oxidizing agents, bases, alcohols and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Chemical Identifiers

CAS 543-20-4
Molecular Formula C4H4Cl2O2
Molecular Weight (g/mol) 154.974
MDL Number MFCD00000749
InChI Key IRXBNHGNHKNOJI-UHFFFAOYSA-N
Synonym succinyl chloride, succinyl dichloride, succinic acid dichloride, succinoyl chloride, succinoyl dichloride, succinic chloride, 1,2-bis chlorocarbonyl ethane, butanedioyl chloride, unii-gdn09v9867, 1,2-ethanediylbis chloroformate
PubChem CID 10970
IUPAC Name butanedioyl dichloride
SMILES C(CC(=O)Cl)C(=O)Cl

Specifications

Melting Point 16°C to 17°C
Density 1.407
Boiling Point 190°C
Flash Point 76°C (168°F)
Odor Acrid
Refractive Index 1.471
Quantity 50 g
UN Number UN3265
Beilstein 507064
Sensitivity Moisture sensitive
Merck Index 14,8873
Solubility Information Reacts violently with water.
Formula Weight 154.98
Percent Purity 96%
Chemical Name or Material Succinyl chloride
Show More Show Less
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 208-838-0
RTECSNumber WN4900000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only