missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Succinimidyl acetate

Catalog No. AC464750050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464750050 5 g
AC464750250 25 g
AC464751000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC464750050 Supplier Thermo Scientific Chemicals Supplier No. 464750050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 14464-29-0 | C6H7NO4 | 157.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14464-29-0
Molecular Formula C6H7NO4
Molecular Weight (g/mol) 157.13
InChI Key SIFCHNIAAPMMKG-UHFFFAOYSA-N
IUPAC Name 2,5-dioxopyrrolidin-1-yl acetate
SMILES CC(=O)ON1C(=O)CCC1=O

Specifications

Melting Point 133.0°C to 134.0°C
Quantity 5 g
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in toluene
Formula Weight 157.13
Chemical Name or Material Succinimidyl acetate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation.

Causes serious eye irritation.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

If skin irritation occurs: Get medical advice/attention.

If eye irritation persists: Get medical advice/attention.

Wear protective gloves/protective clothing/eye protection/face protection.

EINECSNumber 238-454-9

RUO – Research Use Only