missing translation for 'onlineSavingsMsg'
Learn More

Succinimidyl 6-[[7-(N,N-Dimethylaminosulfonyl)-2,1,3-benzoxadiazol-4-yl]amino]hexanoate 98.0+%, TCI America™

Catalog No. S0503100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0503100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S0503100MG Supplier TCI America Supplier No. S0503100MG

Chemical Identifiers

CAS 1858255-08-9
Molecular Formula C18H23N5O7S
Molecular Weight (g/mol) 453.47
InChI Key MMPAJYBUTABRFE-UHFFFAOYSA-N
Synonym 4-(N,N-Dimethylaminosulfonyl)-7-[5-(succinimidyloxycarbonyl)pentylamino]-2,1,3-benzoxadiazole
PubChem CID 44629781
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 6-[[4-(dimethylsulfamoyl)-2,1,3-benzoxadiazol-7-yl]amino]hexanoate
SMILES CN(C)S(=O)(=O)C1=CC=C(C2=NON=C12)NCCCCCC(=O)ON3C(=O)CCC3=O

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 453.47
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Succinimidyl 6-[[7-(N,N-Dimethylaminosulfonyl)-2,1,3-benzoxadiazol-4-yl]amino]hexanoate
TSCA No
Recommended Storage Refrigerator