missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Succinic Dihydrazide 97.0+%, TCI America™
Supplier: TCI America S0482100G
Specifications
| Succinic Dihydrazide | |
| 168°C | |
| C4H10N4O2 | |
| 100 g | |
| HCOMFAYPHBFMKU-UHFFFAOYSA-N | |
| butanedihydrazide | |
| 72491 | |
| ≥97.0% (HPLC,T) |
| 4146-43-4 | |
| White-Yellow | |
| MFCD00007613 | |
| Butanediohydrazide | |
| NNC(=O)CCC(=O)NN | |
| 146.15 | |
| 146.15 | |
| Crystalline Powder |
Chemical Identifiers
| 4146-43-4 | |
| 146.15 | |
| HCOMFAYPHBFMKU-UHFFFAOYSA-N | |
| 72491 | |
| NNC(=O)CCC(=O)NN |
| C4H10N4O2 | |
| MFCD00007613 | |
| Butanediohydrazide | |
| butanedihydrazide |
Safety and Handling
EINECSNumber : (2)-0902
RTECSNumber : WM7500000
TSCA : No