missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Succinic Anhydride 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 120°C |
| Boiling Point | 131°C |
| Color | White |
| Formula Weight | 100.07 |
| Percent Purity | ≥95.0% (T) |
| Quantity | 500 g |
| Physical Form | Pellets |
| Chemical Name or Material | Succinic Anhydride |
Safety and Handling
| EINECSNumber | (2)-0921 |
| RTECSNumber | WN0875000 |
| TSCA | Yes |