missing translation for 'onlineSavingsMsg'
Learn More

Succinic Anhydride 95.0+%, TCI America™

Catalog No. S010725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S010725G 25 g
S0107500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S010725G Supplier TCI America Supplier No. S010725G

Chemical Identifiers

CAS 108-30-5
Molecular Formula C4H4O3
Molecular Weight (g/mol) 100.07
MDL Number MFCD00005525
InChI Key RINCXYDBBGOEEQ-UHFFFAOYSA-N
Synonym succinic anhydride, dihydrofuran-2,5-dione, succinic acid anhydride, succinyl oxide, 2,5-diketotetrahydrofuran, dihydro-2,5-furandione, butanedioic anhydride, 2,5-furandione, dihydro, succinyl anhydride, rikacid sa
PubChem CID 7922
ChEBI CHEBI:36595
IUPAC Name oxolane-2,5-dione
SMILES O=C1CCC(=O)O1

Specifications

Melting Point 120°C
Boiling Point 131°C
Color White
Formula Weight 100.07
Percent Purity ≥95.0% (T)
Quantity 25 g
Physical Form Pellets
Chemical Name or Material Succinic Anhydride
EINECSNumber (2)-0921
RTECSNumber WN0875000
TSCA Yes