missing translation for 'onlineSavingsMsg'
Learn More

Succinic anhydride, MilliporeSigma™

For synthesis

Supplier:  Technidata 8006830100

Catalog No. M8006830100


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2.7 (20g/L, aq.)
116°C to 120°C
Colorless
261°C
Stinging
Crystals
MFCD00005525
Water: 67g/L at 20°C Hydrolysis
O=C1CCC(=O)O1
100.07
CHEBI:36595
Succinic anhydride
108-30-5
1.48g/cm3
100 g
Plastic Bottle
C4H4O3
succinic anhydride, dihydrofuran-2,5-dione, succinic acid anhydride, succinyl oxide, 2,5-diketotetrahydrofuran, dihydro-2,5-furandione, butanedioic anhydride, 2,5-furandione, dihydro, succinyl anhydride, rikacid sa
RINCXYDBBGOEEQ-UHFFFAOYSA-N
oxolane-2,5-dione
7922
100.07

Chemical Identifiers

108-30-5
100.07
RINCXYDBBGOEEQ-UHFFFAOYSA-N
7922
oxolane-2,5-dione
C4H4O3
MFCD00005525
succinic anhydride, dihydrofuran-2,5-dione, succinic acid anhydride, succinyl oxide, 2,5-diketotetrahydrofuran, dihydro-2,5-furandione, butanedioic anhydride, 2,5-furandione, dihydro, succinyl anhydride, rikacid sa
CHEBI:36595
O=C1CCC(=O)O1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H319: Causes serious eye irritation.

H335: May cause respiratory irritation

H302: Harmful if swallowed.

EINECSNumber : 203-570-0

RTECSNumber : WN0875000

Recommended Storage : 15° to 25°C (59° to 77°F); Tightly closed. Dry.

SDS
Documents

Documents

Promotions

Promotions