missing translation for 'onlineSavingsMsg'
Learn More

Succinic anhydride, 99%

Catalog No. AAA1224530
Change view
Click to view available options
Quantity:
250 g
500 g
1000 g
5000 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1224530 250 g
AAA1224536 500 g
AAA122450B 1000 g
AAA122450I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1224530 Supplier Thermo Scientific Chemicals Supplier No. A1224530
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108-30-5 | C4H4O3 | 100.07 g/mol

Succinic anhydride is used as a sizing agent and curative for epoxy resin. It acts as an additive in paper production. It is utilized in the preparation of covalently cross-linked oxidized-alginate and N-succinyl-chitosan hydrogels. It serves as an intermediate for functionalized oxide surfaces on a chip and for polyester resins. Further, it finds use in cosmetics, pharmaceuticals and in coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Succinic anhydride is used as a sizing agent and curative for epoxy resin. It acts as an additive in paper production. It is utilized in the preparation of covalently cross-linked oxidized-alginate and N-succinyl-chitosan hydrogels. It serves as an intermediate for functionalized oxide surfaces on a chip and for polyester resins. Further, it finds use in cosmetics, pharmaceuticals and in coatings.

Solubility
Soluble in chloroform.Soluble in chloroform, carbon tetrachloride and alcohol. Slightly soluble in ether. Insoluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, alcohols and amines.

Chemical Identifiers

CAS 108-30-5
Molecular Formula C4H4O3
Molecular Weight (g/mol) 100.07
MDL Number MFCD00005525
InChI Key RINCXYDBBGOEEQ-UHFFFAOYSA-N
Synonym succinic anhydride, dihydrofuran-2,5-dione, succinic acid anhydride, succinyl oxide, 2,5-diketotetrahydrofuran, dihydro-2,5-furandione, butanedioic anhydride, 2,5-furandione, dihydro, succinyl anhydride, rikacid sa
PubChem CID 7922
ChEBI CHEBI:36595
IUPAC Name oxolane-2,5-dione
SMILES O=C1CCC(=O)O1

Specifications

Melting Point 118°C to 121°C
Boiling Point 261°C
Density 1.572
Quantity 250 g
Flash Point 157°C (314°F)
Assay Percent Range 0.99
Sensitivity Moisture sensitive
Beilstein 108441
Merck Index 148870
Solubility Information Soluble in chloroform;Soluble in chloroform,carbon tetrachloride and alcohol; Slightly soluble in ether; Insoluble in water.
Formula Weight 100.07
Percent Purity 99%
Chemical Name or Material Succinic Anhydride
Show More Show Less
Hazard Category H302-H314-H317-H334
Hazard Statement GHS H Statement
H302-H319-H335
Precautionary Statement P260-P264b-P270-P271-P272-P280-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 203-570-0
RTECSNumber WN0875000
TSCA Yes
storageNote1 Moisture Sensitive
Recommended Storage Ambient temperatures

RUO – Research Use Only