missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Succinic Acid 99.0+%, TCI America™
Supplier: TCI America S010025G
Specifications
| Succinic Acid | |
| 187°C | |
| 235°C | |
| C4H6O4 | |
| succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid | |
| OC(=O)CCC(O)=O | |
| 118.09 | |
| CHEBI:15741 | |
| ≥99.0% (T) | |
| Crystalline Powder |
| 2.7 (0.1M H2O soln.) | |
| 110-15-6 | |
| White | |
| MFCD00002789 | |
| KDYFGRWQOYBRFD-UHFFFAOYSA-N | |
| butanedioic acid | |
| 1110 | |
| 118.09 | |
| 25 g |
Chemical Identifiers
| 110-15-6 | |
| 118.09 | |
| KDYFGRWQOYBRFD-UHFFFAOYSA-N | |
| 1110 | |
| butanedioic acid |
| C4H6O4 | |
| MFCD00002789 | |
| succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid | |
| CHEBI:15741 | |
| OC(=O)CCC(O)=O |
Safety and Handling
EINECSNumber : (2)-0846
RTECSNumber : WM4900000
TSCA : Yes