missing translation for 'onlineSavingsMsg'
Learn More

Succinic Acid, FCC, 99-100.5%, Spectrum™ Chemical

Catalog No. 18607051
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607051 2.5 kg Poly Pail
18607050 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18607051 Supplier Spectrum Chemical Mfg Cor Supplier No. S16862.5KG
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

S1686, 110-15-6, C4H6O4

Spectrum™ Chemical Succinic Acid, FCC is used as a flavoring agent for food and beverages. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 110-15-6
Molecular Formula C4H6O4
Molecular Weight (g/mol) 118.09
MDL Number MFCD00002789
InChI Key KDYFGRWQOYBRFD-UHFFFAOYSA-N
IUPAC Name butanedioic acid
SMILES OC(=O)CCC(O)=O

Specifications

CAS Max % 1
Quantity 2.5 kg
Ignition Residue 0.00025
Packaging Poly Pail
Percent Purity 99.0 to 100.5%
Grade FCC