missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Succinic acid disodium salt, hexahydrate, >98%, MP Biomedicals™
$58.05 - $58.05
Chemical Identifiers
| CAS | 150-90-3 |
|---|---|
| Molecular Formula | C4H4Na2O4 |
| Molecular Weight (g/mol) | 162.052 |
| InChI Key | ZDQYSKICYIVCPN-UHFFFAOYSA-L |
| Synonym | disodium succinate, sodium succinate, disodium butanedioate, butanedioic acid, disodium salt, soduxin, succinic acid disodium salt, jantaran sodny czech, unii-v8zgc8isr3, succinic acid, disodium salt, fema no. 3277 |
| PubChem CID | 9020 |
| ChEBI | CHEBI:63675 |
| IUPAC Name | disodium;butanedioate |
| SMILES | C(CC(=O)[O-])C(=O)[O-].[Na+].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN10292980
|
MP Biomedicals Inc
0210292980 |
100 g |
Each for $58.05
|
|
|||||
Chemical Identifiers
| 150-90-3 | |
| 162.052 | |
| disodium succinate, sodium succinate, disodium butanedioate, butanedioic acid, disodium salt, soduxin, succinic acid disodium salt, jantaran sodny czech, unii-v8zgc8isr3, succinic acid, disodium salt, fema no. 3277 | |
| CHEBI:63675 | |
| C(CC(=O)[O-])C(=O)[O-].[Na+].[Na+] |
| C4H4Na2O4 | |
| ZDQYSKICYIVCPN-UHFFFAOYSA-L | |
| 9020 | |
| disodium;butanedioate |
Specifications
| 150-90-3 | |
| C4H4Na2O4 | |
| disodium succinate, sodium succinate, disodium butanedioate, butanedioic acid, disodium salt, soduxin, succinic acid disodium salt, jantaran sodny czech, unii-v8zgc8isr3, succinic acid, disodium salt, fema no. 3277 | |
| C(CC(=O)[O-])C(=O)[O-].[Na+].[Na+] | |
| 162.052 | |
| CHEBI:63675 | |
| >98% | |
| Succinic acid disodium salt, hexahydrate |
| White | |
| 100 g | |
| ZDQYSKICYIVCPN-UHFFFAOYSA-L | |
| disodium;butanedioate | |
| 9020 | |
| 270.1 | |
| Powder |