missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Succinic acid, disodium salt hexahydrate, 97%
CAS: 6106-21-4 | C4H16Na2O10 | 270.142 g/mol
$169.54 - $169.54
Chemical Identifiers
| CAS | 6106-21-4 |
|---|---|
| Molecular Formula | C4H16Na2O10 |
| Molecular Weight (g/mol) | 270.142 |
| InChI Key | ZBTUYCUNQBRXOR-UHFFFAOYSA-L |
| Synonym | disodium succinate hexahydrate, sodium succinate hexahydrate, unii-u16qod6c4e, butanedioic acid, disodium salt, hexahydrate, disodium butanedioate hexahydrate, u16qod6c4e, disodium hexahydrate succinate, sodium succinate dibasic hexahydrate, butanedioic acid disodium salt, succinic acid disodium salt hexahydrate |
| PubChem CID | 3083938 |
| ChEBI | CHEBI:63686 |
| IUPAC Name | disodium;butanedioate;hexahydrate |
| SMILES | C(CC(=O)[O-])C(=O)[O-].O.O.O.O.O.O.[Na+].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC158755000
|
Thermo Scientific Chemicals
158755000 |
500 g |
Each for $169.54
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 6106-21-4 | |
| 270.142 | |
| disodium succinate hexahydrate, sodium succinate hexahydrate, unii-u16qod6c4e, butanedioic acid, disodium salt, hexahydrate, disodium butanedioate hexahydrate, u16qod6c4e, disodium hexahydrate succinate, sodium succinate dibasic hexahydrate, butanedioic acid disodium salt, succinic acid disodium salt hexahydrate | |
| CHEBI:63686 | |
| C(CC(=O)[O-])C(=O)[O-].O.O.O.O.O.O.[Na+].[Na+] |
| C4H16Na2O10 | |
| ZBTUYCUNQBRXOR-UHFFFAOYSA-L | |
| 3083938 | |
| disodium;butanedioate;hexahydrate |
Specifications
| 6106-21-4 | |
| Brown to Yellow | |
| 37.0 % to 41.0 % (90°C) | |
| 0.97 | |
| C4H16Na2O10 | |
| 02, 606 | |
| disodium succinate hexahydrate, sodium succinate hexahydrate, unii-u16qod6c4e, butanedioic acid, disodium salt, hexahydrate, disodium butanedioate hexahydrate, u16qod6c4e, disodium hexahydrate succinate, sodium succinate dibasic hexahydrate, butanedioic acid disodium salt, succinic acid disodium salt hexahydrate | |
| ZBTUYCUNQBRXOR-UHFFFAOYSA-L | |
| disodium;butanedioate;hexahydrate | |
| 3083938 | |
| 270.13 | |
| 0.005% max. | |
| Succinic acid, disodium salt hexahydrate, 0.97 |
| 120.0°C | |
| 7 to 9 (0.4M soln.) | |
| Authentic | |
| Glass bottle | |
| 500 g | |
| 15, 8808 | |
| Solubility in water: 270g/L water (25°C). Other solubilities: insoluble in alcohol | |
| C(CC(=O)[O-])C(=O)[O-].O.O.O.O.O.O.[Na+].[Na+] | |
| 270.142 | |
| CHEBI:63686 | |
| 0.97 | |
| Liquid |
RUO – Research Use Only