missing translation for 'onlineSavingsMsg'
Learn More

Succinic Acid, Disodium Salt, 6-Hydrate, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. p-5391503
Change view
Click to view available options
Quantity:
250 g
5 kg
Packaging:
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
02004392 250 g Poly Bottle
02004393 5 kg Poly Pail
2 options

Catalog No. 02004392

Supplier: Avantor V10306

Only null left

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 6106-21-4
Molecular Formula C4H16Na2O10
Molecular Weight (g/mol) 270.142
InChI Key ZBTUYCUNQBRXOR-UHFFFAOYSA-L
Synonym disodium succinate hexahydrate, sodium succinate hexahydrate, unii-u16qod6c4e, butanedioic acid, disodium salt, hexahydrate, disodium butanedioate hexahydrate, u16qod6c4e, disodium hexahydrate succinate, sodium succinate dibasic hexahydrate, butanedioic acid disodium salt, succinic acid disodium salt hexahydrate
PubChem CID 3083938
ChEBI CHEBI:63686
IUPAC Name disodium;butanedioate;hexahydrate
SMILES C(CC(=O)[O-])C(=O)[O-].O.O.O.O.O.O.[Na+].[Na+]

Specifications

Packaging Poly Bottle
Quantity 250 g
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Succinic Acid, Disodium Salt, 6-Hydrate