missing translation for 'onlineSavingsMsg'
Learn More

Succinic Acid, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical

Catalog No. 18607042
Change view
Click to view available options
Quantity:
125 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607042 125 g Poly Bottle
18607043 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18607042 Supplier Spectrum Chemical Mfg Cor Supplier No. S1685125GM
Get Up to $500 in Chemicals with Purchase
View all promotions

S1685, 110-15-6, C4H6O4

Spectrum™ Chemical Succinic Acid, Crystal, Reagent, ACS is a white, odorless solid that is a dicarboxylic acid. Advances in industrial biotechnology that seek to displace petroleum-based chemicals in industrial applications are fueling growth in this product. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 110-15-6
Molecular Formula C4H6O4
Molecular Weight (g/mol) 118.09
MDL Number MFCD00002789
InChI Key KDYFGRWQOYBRFD-UHFFFAOYSA-N
IUPAC Name butanedioic acid
SMILES OC(=O)CCC(O)=O

Specifications

Melting Point 185°C to 191°C
CAS Max % 1
Percent Purity 99%
Grade ACS
Quantity 125 g
Packaging Poly Bottle
Residue after Ignition 0.0002