missing translation for 'onlineSavingsMsg'
Learn More

Succinamic acid, 97%

Catalog No. AC158731000
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC158731000 100 g Glass bottle
AC158730050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC158731000 Supplier Thermo Scientific Chemicals Supplier No. 158731000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 638-32-4 | C4H7NO3 | 117.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 638-32-4
Molecular Formula C4H7NO3
Molecular Weight (g/mol) 117.10
MDL Number MFCD00008041
InChI Key JDVPQXZIJDEHAN-UHFFFAOYSA-N
Synonym succinamic acid, butanoic acid, 4-amino-4-oxo, succinic acid monoamide, 3-carbamoylpropanoic acid, succinamidic acid, acmc-1b64y, 3-c-hydroxycarbonimidoyl propanoic acid
PubChem CID 12522
ChEBI CHEBI:50398
IUPAC Name 4-amino-4-oxobutanoic acid
SMILES NC(=O)CCC(O)=O

Specifications

Melting Point 153°C to 157°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula H2NCOCH2CH2CO2H
Quantity 100 g
Beilstein 02, 614
Formula Weight 117.1
Percent Purity 97%
Physical Form Crystalline Powder or Needles
Chemical Name or Material Succinamic acid, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-331-7

RUO – Research Use Only