Learn More
Abcam Inc Suc-Leu-Leu-Val-Tyr-AMC, Fluorogenic substrate of calpains and 20S proteasome, 10MG
Catalog No. 502706326
Encompass
MW 763.9 Da, Purity >97%. Suc-Leu-Leu-Val-Tyr-AMC, Fluorogenic substrate of calpains and 20S proteasome.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 502706326 Supplier Abcam Inc Supplier No. AB14212010MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MW 763.9 Da, Purity >97%. Suc-Leu-Leu-Val-Tyr-AMC, Fluorogenic substrate of calpains and 20S proteasome.
The product is subject to the following: Abcam Restricted Use Statement