Learn More
Substance P
For cellular and molecular biology applications | 1347.64 g/mol
Supplier: Thermo Scientific Chemicals J63489LB0
Description
Substance P is used as a neurokinin NK-1 receptor agonist, neurokinin undecapeptide neurotransmitter and neuromodulator, also used in cancer cell proliferation and that inhibition of this component can induce apoptosis in cancer cells and decrease the steady state of Her2.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| HPLC | |
| A neurokinin NK-1 receptor agonist | |
| Substance P | |
| RUO - research use only |
| Powder | |
| 1347.64 | |
| 5 mg | |
| Arg-Pro-Lys-Pro-Gln-Gln-Phe-Phe-Gly-Leu-Met-NH2 |
RUO – Research Use Only