missing translation for 'onlineSavingsMsg'
Learn More

Substance P

Catalog No. AAJ63489MB
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63489MB 25 mg
AAJ63489LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63489MB Supplier Thermo Scientific Chemicals Supplier No. J63489MB

For cellular and molecular biology applications | 1347.64 g/mol

Substance P is used as a neurokinin NK-1 receptor agonist, neurokinin undecapeptide neurotransmitter and neuromodulator, also used in cancer cell proliferation and that inhibition of this component can induce apoptosis in cancer cells and decrease the steady state of Her2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Purification Method HPLC
Format Powder
For Use With (Application) A neurokinin NK-1 receptor agonist
Molecular Weight (g/mol) 1347.64
Product Type Substance P
Quantity 25 mg
Regulatory Status RUO
Sequence Arg-Pro-Lys-Pro-Gln-Gln-Phe-Phe-Gly-Leu-Met-NH2

RUO – Research Use Only