Learn More
Chemglass Life Sciences Micro Sublimer, Complete, 24/25 Jts., 160mm Height x 85mm Width
Supplier: Chemglass Inc CG303001
Micro Sublimer, Complete, 24/25 Jts., 160mm Height x 85mm Width. Micro scale sublimer can be used with quantities as small as 0.5 grams. The sublimer has a capacity of 3mL between the lower portion of the cold finger and the interior bottom of the flask. With 24/25 standard taper joints to connect the flask and cold finger condenser.