missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sublimed Sulfur, 99.5-100.5%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Physical Form | Crystalline Powder or Needles |
| Quantity | 45 kg |
| Molecular Weight (g/mol) | 32.07 |
| Percent Purity | 99.5 to 100.5% |
| Grade | Not Available |
| Identification | To Pass Test |
| Packaging | Poly Pail |
| Chemical Name or Material | Sublimed Sulfur |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.