missing translation for 'onlineSavingsMsg'
Learn More

Suberoyl-L-Carnitine-(N-Methyl-D3) Lithium Salt Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022806
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022806 10 mg
111022808 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111022806 Supplier Merck Emd Millipore Supplier No. 9258610MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 1803252-74-5 (Free Acid)
Molecular Formula C15D3H24NO6 · xLi+
Linear Formula C15D3H24NO6 · xLi+
Quantity 10 mg
Optical Rotation [α]/D -17°C ± 2°C = 0.1 in. H2O
Formula Weight 320.40 (Free Acid Basis)
Percent Purity ≥95% (HPLC)
Grade Analytical Standard