missing translation for 'onlineSavingsMsg'
Learn More

Suberoyl Chloride 98.0+%, TCI America™

Catalog No. S042025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S042025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S042025G Supplier TCI America Supplier No. S042025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10027-07-3
Molecular Formula C8H12Cl2O2
Molecular Weight (g/mol) 211.082
MDL Number MFCD00000764
InChI Key PUIBKAHUQOOLSW-UHFFFAOYSA-N
Synonym suberoyl chloride, suberoyl dichloride, suberic acid dichloride, suberyl chloride, suberyl dichloride, acmc-1bonz, octanedioic acid dichloride
PubChem CID 534653
IUPAC Name octanedioyl dichloride
SMILES C(CCCC(=O)Cl)CCC(=O)Cl

Specifications

Color Yellow
Boiling Point 147°C
Quantity 25 g
UN Number 3265
Formula Weight 211.08
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Suberoyl Chloride