missing translation for 'onlineSavingsMsg'
Learn More

Suberoyl chloride, 97%

Catalog No. AAB2458006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2458006 5 g
AAB2458014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2458006 Supplier Thermo Scientific Chemicals Supplier No. B2458006

CAS: 10027-07-3 | C8H12Cl2O2 | 211.082 g/mol

Suberoyl chloride is used as a reagent to synthesize hydroxyferrocifen hybrid compounds that have antiproliferative activity against triple negative breast cancer cells. It is also used as a cross-linking agent to cross-link Chitosan (C315015) membranes, and also improves the membrane’s integrity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Suberoyl chloride is used as a reagent to synthesize hydroxyferrocifen hybrid compounds that have antiproliferative activity against triple negative breast cancer cells. It is also used as a cross-linking agent to cross-link Chitosan (C315015) membranes, and also improves the membrane’s integrity.

Solubility
Reacts with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is sensitive to moisture. Store under dry inert gas. Protect from humidity and water.

Chemical Identifiers

CAS 10027-07-3
Molecular Formula C8H12Cl2O2
Molecular Weight (g/mol) 211.082
MDL Number MFCD00000764
InChI Key PUIBKAHUQOOLSW-UHFFFAOYSA-N
Synonym suberoyl chloride, suberoyl dichloride, suberic acid dichloride, suberyl chloride, suberyl dichloride, acmc-1bonz, octanedioic acid dichloride
PubChem CID 534653
IUPAC Name octanedioyl dichloride
SMILES C(CCCC(=O)Cl)CCC(=O)Cl

Specifications

Density 1.172
Boiling Point 162°C to 163°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.468
Linear Formula ClOC(CH2)6COCl
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 211.09
Percent Purity 97%
Chemical Name or Material Suberoyl chloride
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only