missing translation for 'onlineSavingsMsg'
Learn More

Suberonitrile 98.0+%, TCI America™

Catalog No. p-7135858
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S029825G 25 g
S0298250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S029825G Supplier TCI America Supplier No. S029825G
Only null left

Chemical Identifiers

CAS 629-40-3
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.198
MDL Number MFCD00001981
InChI Key BTNXBLUGMAMSSH-UHFFFAOYSA-N
Synonym 1,6-dicyanohexane, suberonitrile, suberic acid dinitrile, suberodinitrile, unii-jnt1lxa02u, korksaeuredinitril german, jnt1lxa02u, hexamethylene di-isocyanide, hexane-1,6-dicarbonitrile, hexamethylenediisocyanic acid
PubChem CID 12385
IUPAC Name octanedinitrile
SMILES C(CCCC#N)CCC#N

Specifications

Color Colorless
Boiling Point 172°C
Quantity 25 g
UN Number 3276
Formula Weight 136.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Suberonitrile
EINECSNumber (2)-3619
RTECSNumber WM2131000
TSCA Yes