missing translation for 'onlineSavingsMsg'
Learn More

Suberic Acid 99.0+%, TCI America™

Catalog No. O002325G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O002325G 25 g
O0023100G 100 g
O0023500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O002325G Supplier TCI America Supplier No. O002325G

Chemical Identifiers

CAS 505-48-6
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.20
MDL Number MFCD00004428
InChI Key TYFQFVWCELRYAO-UHFFFAOYSA-N
Synonym suberic acid, cork acid, 1,6-hexanedicarboxylic acid, 1,8-octanedioic acid, hexamethylenedicarboxylic acid, subericacid, 1,6-dicarboxyhexane, octane-1,8-dioic acid, octanedioc acid, unii-6u7y4m9c1h
PubChem CID 10457
ChEBI CHEBI:9300
IUPAC Name octanedioic acid
SMILES OC(=O)CCCCCCC(O)=O

Specifications

Melting Point 143°C
Color White
Boiling Point 279°C
Quantity 25 g
Formula Weight 174.20
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Suberic Acid
EINECSNumber (2)-3505
TSCA Yes