missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Suberic Acid 99.0+%, TCI America™
Supplier: TCI America O0023100G
| Quantity | 100 g |
|---|
Chemical Identifiers
| C8H14O4 | |
| MFCD00004428 | |
| suberic acid, cork acid, 1,6-hexanedicarboxylic acid, 1,8-octanedioic acid, hexamethylenedicarboxylic acid, subericacid, 1,6-dicarboxyhexane, octane-1,8-dioic acid, octanedioc acid, unii-6u7y4m9c1h | |
| CHEBI:9300 | |
| OC(=O)CCCCCCC(O)=O |
Specifications
| Suberic Acid | |
| 143°C | |
| 279°C | |
| 100 g | |
| suberic acid, cork acid, 1,6-hexanedicarboxylic acid, 1,8-octanedioic acid, hexamethylenedicarboxylic acid, subericacid, 1,6-dicarboxyhexane, octane-1,8-dioic acid, octanedioc acid, unii-6u7y4m9c1h | |
| OC(=O)CCCCCCC(O)=O | |
| 174.20 | |
| CHEBI:9300 | |
| ≥99.0% (GC,T) |
| 505-48-6 | |
| White | |
| C8H14O4 | |
| MFCD00004428 | |
| TYFQFVWCELRYAO-UHFFFAOYSA-N | |
| octanedioic acid | |
| 10457 | |
| 174.20 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-3505
TSCA : Yes