missing translation for 'onlineSavingsMsg'
Learn More

Styrene Oxide 98.0+%, TCI America™

Catalog No. E001325ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E001325ML 25 mL
E0013500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E001325ML Supplier TCI America Supplier No. E001325ML

Chemical Identifiers

CAS 96-09-3
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.151
MDL Number MFCD00005121
InChI Key AWMVMTVKBNGEAK-UHFFFAOYSA-N
Synonym styrene oxide, phenyloxirane, 1,2-epoxyethylbenzene, phenylethylene oxide, epoxystyrene, styrene epoxide, styryl oxide, oxirane, phenyl, 1-phenyloxirane, phenethylene oxide
PubChem CID 7276
ChEBI CHEBI:17907
IUPAC Name 2-phenyloxirane
SMILES C1C(O1)C2=CC=CC=C2

Specifications

Melting Point -37°C
Color Yellow
Boiling Point 194°C
Quantity 25 mL
UN Number 2810
Formula Weight 120.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Styrene Oxide
EINECSNumber (3)-1033
RTECSNumber CZ9625000
TSCA Yes