missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Styrene oxide, 98+%

Catalog No. AAL0782136
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0782136 500 g
AAL0782122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0782136 Supplier Thermo Scientific Chemicals Supplier No. L0782136
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 96-09-3 | C8H8O | 120.151 g/mol

Styrene oxide is used as an intermediate in medicine and perfume.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Styrene oxide is used as an intermediate in medicine and perfume.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 96-09-3
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.151
MDL Number MFCD00005121
InChI Key AWMVMTVKBNGEAK-UHFFFAOYSA-N
Synonym styrene oxide, phenyloxirane, 1,2-epoxyethylbenzene, phenylethylene oxide, epoxystyrene, styrene epoxide, styryl oxide, oxirane, phenyl, 1-phenyloxirane, phenethylene oxide
PubChem CID 7276
ChEBI CHEBI:17907
IUPAC Name 2-phenyloxirane
SMILES C1C(O1)C2=CC=CC=C2

Specifications

Melting Point -37°C
Density 1.053
Boiling Point 193°C to 196°C
Flash Point 74°C (165°F)
Refractive Index 1.535
Quantity 500 g
UN Number UN2810
Beilstein 108582
Solubility Information Soluble in water.
Formula Weight 120.15
Percent Purity ≥98%
Chemical Name or Material (+/-)-Styrene oxide
Show More Show Less
Hazard Category H227-H312-H315-H317-H319-H331-H340-H350
Hazard Statement GHS H Statement
H350-H302-H312-H319-H227
May cause cancer.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P201-P202-P210-P235-P261-P264b-P271-P272-P280-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P312-P333+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 202-476-7
RTECSNumber CZ9625000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only