missing translation for 'onlineSavingsMsg'
Learn More

Carson™ Stuff-It Microfiber Lens Cleaning System

Catalog No. s81708
Encompass
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S81708 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S81708 Supplier Carson™ Supplier No. SN40BK

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Stuff-It from Carson is a 6.25 x 6.25 in. microfiber cleaning cloth that is housed inside of a protective neoprene pouch. The Stuff-It also has a carabiner clip attached by a drawstring so users can easily attach it to keychains, backpacks, belt loops, and more.

  • The Stuff-It microfiber lens cleaning system is a safe, easy and effective way to clean all types of lenses, screens, and glass surfaces.
  • To use, simply pull the cloth out of the protective pouch.
  • After you're done using the cloth, simply use your fingers to stuff the cloth back into the pouch.
  • The Stuff-It will not scratch or damage surfaces in any way.
  • It is a great cleaning device for smartphones, tablets, eyeglasses, cameras, optics, and so much more.
  • Note: Hand Wash Only

Specifications

Product Type Cleaning Cloth
Material Microfiber
Absorbancy Yes
Autoclavable Not Autoclavable
Basis Weight 0.034 lb.
Color Black
Disposable Reusable
Edges Chiseled
Dimensions (L x W) Cloth: 6.5 x 6.5 in.; Protective Pouch: 1.7 x 1.1 in.
For Use With (Application) Cleaning Smartphones, Eyeglasses, Binoculars, Scopes, Optics, and More
Sterility Non-sterile
Includes Microfiber Cleaning Cloth with Built-in Protective Pouch
Show More Show Less