missing translation for 'onlineSavingsMsg'
Learn More
Learn More
United Scientific™ Student Plasticware Kit
Description
Includes
- Two cylinders
- Two beakers
- Three test tubes
- Four bottles
- Six dropping pipets
- One funnel
- One dispenser
- One wash bottle
- Kit with plastic storage box consists of 20 items.
Specifications
Specifications
| Grade | 5+ |
| Includes | One 100 mL beaker, one 250 mL beaker, one 10 mL graduated cylinder, one 100 mL graduated cylinder, 1 funnel, 65 mm top dia., 1 drop dispenser bottle, 4 reagent bottle, 1 wash bottle, 3 plastic test tubes, 6 pipettes |
| Class Size | 1 to 2 Lab Groups |
| Product Type | Plasticware Kit |