missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Student Optics Kit and Accessories

The Student Optics Kit is an user-friendly tool for students to learn about optics in a visually and kinesthetically effective manner. The kit includes color cards, slit cards, color filters, blank plates, acrylic optical shapes, and tray with measurement markings

$23.21 - $405.64

Specifications

For Use With (Application) Optics Education
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
S27720
View Documents
Eisco™
PH0615A
Kit, Student Optics
Each for $405.64
Only null left
 
S27721
View Documents
Eisco™
PH0615AD
Light Filter Set
Each for $23.21
Only null left
 
Description

Description

  • Color Card, Color Filter, and Black Plastic Blanks Dimensions: 2 x 2 in.
  • Slit Card Dimensions: 2 x 3.38 in.
  • 6 Acrylic Optical Bodies of various shapes
  • Slots in ray box for placement of cards, filters, etc.
Specifications

Specifications

Optics Education
Videos
SDS
Documents

Documents