missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Student Optical Bench

Catalog No. S80854
Change view
Click to view available options
Student Optical Bench
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S80854 Student Optical Bench
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S80854 Supplier Eisco™ Supplier No. PH0647

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A cost effective way to demonstrate the basic principles of optics in a typical high school science lab

  • Cost effective student optical bench
  • Demonstrates basic optical principles
  • Economy model

Specifications

Product Type Student Optical Bench
Grade 6 to Undergraduate
For Use With (Application) Demonstration of Basic Optical Principals
Includes Meter Stick, 2 Meter Stick Supports, Object Marker, Screen Supports, Card with Screen, and Lens Support (38 mm)