missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Student Magnetic Field Demonstrator

Catalog No. S16077
Change view
Click to view available options
Grade:
5+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S16077 5+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16077 Supplier United Scientific™ Supplier No. MGFD01*

Economical and effective way to demonstrate magnetic fields

  • Iron fillings, sealed in a transparent plastic case.
  • Measures approximately 90 x 65 x 6 mm

Specifications

Material 100% Iron
For Use With (Application) Magnetic Demonstrations
Grade 5+
Product Type Magnetic Field Demonstrator