Learn More
Student-Grade Tuning Fork Sets
Economical set.
Supplier:  Customer Channels Group (Ccg) 70013STEC
 View more versions of this product
Catalog No. S68769
Only null left
Description
Description
  • Eight-piece set includes forks with 256, 288, 320, 341.1, 384, 426.7, 480 and 512Hz
  • 13-Piece set includes forks with 261.6, 277.1, 293.7, 311.1, 329.6, 349.2, 369.9, 392, 415.30, 440, 466.1, 493.9 and 523.2Hz
Specifications
Chemical Identifiers
Tuning Fork Set
Specifications
Tuning Fork Set
13 Piece set includes forks with 261.6, 277.1, 293.7, 311.1, 329.6, 349.2, 369.9, 392, 415.30, 440, 466.1, 493.9 and 523.2Hz
13/Set
Product Suggestions
Product Suggestions