missing translation for 'onlineSavingsMsg'
Learn More

DR Instruments Student Dissecting Kit with Case

Catalog No. s06831
Change view
Click to view available options
Case Material:
Vinyl
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Case Material
S06831 Vinyl
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

California Proposition 65 WARNING: Cancer and Reproductive Harm

Catalog No. S06831 Supplier DR Instruments Supplier No. 65
Only null left

California Proposition 65 WARNING: Cancer and Reproductive Harm

Complete Student Dissecting Kit with Case for use in the lab or in-class dissection.

Kit with Black Vinyl Case:

  • Dissecting scissors, 4.5 in.
  • Dissecting forceps, 4.5 in. Stainless Steel
  • Dissecting scalpel, Stainless Steel, Super Sharp
  • Two dissecting teasing needles (one angular, one straight) with plastic handles
  • Glass dropper with rubber bulb
  • Plastic ruler, 6 in.
  • Leatherette case with 6 loops

Kit with See-Through Plastic Case:

  • Dissecting scissors, 4.5 in.
  • Dissecting forceps, 4.5 in. chrome-plated
  • Dissecting scalpel, chrome-plated
  • Two dissecting teasing needles (one angular, one straight) with plastic handles
  • Plastic ruler, 6 in.
  • Hard see through top plastic case for storage

Specifications

Quantity 1 Ea.
Case Material Vinyl
For Use With (Application) Complete dissection kits for use in the student lab
Product Type Student Dissecting Kit
Includes Black Vinyl case, 4.5 in. dissecting scissors, 4.5 in. dissecting forceps Chrome-plated, dissecting scalpel, Chrome-plated, two dissecting teasing needles (one angular, one straight) with Plastic handles, Glass dropper with Rubber bulb, 6 in. Plastic ruler