missing translation for 'onlineSavingsMsg'
Learn More

Strychnine Nitrate 99.0+%, TCI America™

Catalog No. S009325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S009325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S009325G Supplier TCI America Supplier No. S009325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 66-32-0
Molecular Formula C21H23N3O5
Molecular Weight (g/mol) 397.431
MDL Number MFCD00078519
InChI Key PCGVPMHGSJFFTI-ZEYGOCRCSA-N
PubChem CID 23616203
IUPAC Name (4aR,5aS,8aR,13aS,15aS,15bR)-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one;nitric acid
SMILES C1CN2CC3=CCOC4CC(=O)N5C6C4C3CC2C61C7=CC=CC=C75.[N+](=O)(O)[O-]

Specifications

Color White
Quantity 25 g
UN Number 1692
Formula Weight 397.43
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Strychnine Nitrate
RTECSNumber WL2450000
TSCA No