missing translation for 'onlineSavingsMsg'
Learn More

Strychnine Hydrochloride, Dihydrate, 98%, Spectrum™ Chemical

Catalog No. p-7166617
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614120 5 g Amber Glass Bottle
18614119 25 g Amber Glass Bottle
18614117 100 g Amber Glass Bottle
18614118 1 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18614120 Supplier Spectrum Chemical Mfg Cor Supplier No. ST1455GM
Only null left

Chemical Identifiers

CAS 6101-04-8

Specifications

CAS Max % 1
Quantity 5 g
Percent Purity 98%
Grade Reagent
Packaging Amber Glass Bottle

CAUTION: For use by qualified personnel for research and development or other authorized use only. Not for food, drug or pesticide use. Read and understand the label and Safety Data Sheet (SDS) prior to use.