missing translation for 'onlineSavingsMsg'
Learn More

Strychnine Hydrochloride, Dihydrate, 98%, Spectrum™ Chemical

Catalog No. 18614118
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614118 1 kg Amber Glass Bottle
18614120 5 g Amber Glass Bottle
18614119 25 g Amber Glass Bottle
18614117 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18614118 Supplier Spectrum Chemical Mfg Cor Supplier No. ST1451KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6101-04-8

Specifications

CAS Max % 1
Physical Form Crystals and/or Chunks
Quantity 1 kg
Percent Purity 98%
Grade Ungraded
Packaging Amber Glass Bottle
Chemical Name or Material Strychnine Hydrochloride, Dihydrate

CAUTION: For use by qualified personnel for research and development or other authorized use only. Not for food, drug or pesticide use. Read and understand the label and Safety Data Sheet (SDS) prior to use.