missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Strontium Titanium Oxide Substrate, 5 x 5 x 0.5mm, Polished One Side, 110 Orientation, Thermo Scientific™
$161.52 - $161.52
Chemical Identifiers
| CAS | 12060-59-2 |
|---|---|
| Molecular Formula | O3SrTi |
| Molecular Weight (g/mol) | 183.48 |
| MDL Number | MFCD00049554 |
| InChI Key | VEALVRVVWBQVSL-UHFFFAOYSA-N |
| Synonym | strontium titanate, strontium titanium oxide, unii-olh4i98373, o3ti.sr, strontium titanium oxide srtio3, strontium titanium trioxide, titanate tio32-, strontium 1:1, strontium titanate 100g, strontium dioxido oxo titanium, strontium titanate, powder |
| PubChem CID | 82899 |
| IUPAC Name | strontium;dioxido(oxo)titanium |
| SMILES | [Sr++].[O-][Ti]([O-])=O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 12060-59-2 | |
| 183.48 | |
| VEALVRVVWBQVSL-UHFFFAOYSA-N | |
| 82899 | |
| [Sr++].[O-][Ti]([O-])=O |
| O3SrTi | |
| MFCD00049554 | |
| strontium titanate, strontium titanium oxide, unii-olh4i98373, o3ti.sr, strontium titanium oxide srtio3, strontium titanium trioxide, titanate tio32-, strontium 1:1, strontium titanate 100g, strontium dioxido oxo titanium, strontium titanate, powder | |
| strontium;dioxido(oxo)titanium |
Specifications
| White | |
| Substrate | |
| O3SrTi | |
| strontium titanate, strontium titanium oxide, unii-olh4i98373, o3ti.sr, strontium titanium oxide srtio3, strontium titanium trioxide, titanate tio32-, strontium 1:1, strontium titanate 100g, strontium dioxido oxo titanium, strontium titanate, powder | |
| [Sr++].[O-][Ti]([O-])=O | |
| 183.48 | |
| Odorless |
| 12060-59-2 | |
| 1 Ea. | |
| MFCD00049554 | |
| VEALVRVVWBQVSL-UHFFFAOYSA-N | |
| strontium;dioxido(oxo)titanium | |
| 82899 | |
| Strontium Titanium Oxide Substrate, Polished One Side |
Safety and Handling
EINECSNumber : 235-044-1
TSCA : Yes
Recommended Storage : Ambient temperatures