missing translation for 'onlineSavingsMsg'
Learn More

Strontium titanium oxide, S.A. 12.5-25m2/g

Catalog No. AA4163422
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4163422 100 g
AA4163436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4163422 Supplier Thermo Scientific Chemicals Supplier No. 04163422
Only null left

CAS: 12060-59-2 | O3SrTi | 183.48 g/mol

Strontium titaniumoxide is used as paint . It has been used as a strontium-90-containing material in radioisotope thermoelectric generators. It is also used as a diamond simulant. This compound is been used in television sets, radios and computers, capacitors, because of the dielectric constant of the material. Used in electronics and in electrical insulation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12060-59-2
Molecular Formula O3SrTi
Molecular Weight (g/mol) 183.48
MDL Number MFCD00049554
InChI Key VEALVRVVWBQVSL-UHFFFAOYSA-N
Synonym strontium titanate, strontium titanium oxide, unii-olh4i98373, o3ti.sr, strontium titanium oxide srtio3, strontium titanium trioxide, titanate tio32-, strontium 1:1, strontium titanate 100g, strontium dioxido oxo titanium, strontium titanate, powder
PubChem CID 82899
IUPAC Name strontium;dioxido(oxo)titanium
SMILES [Sr++].[O-][Ti]([O-])=O

Specifications

Melting Point 2060°C
Physical Form Powder
Quantity 100 g
Solubility Information It is insoluble in water and most solvents.
Formula Weight 183.51
Odor Odorless
Density 4.810 g/mL
Chemical Name or Material Strontium titanium oxide
EINECSNumber 235-044-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only