missing translation for 'onlineSavingsMsg'
Learn More

Strontium sulfate, Puratronic™, 99.9965% (metals basis)

Catalog No. p-7049090
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1287006 5 g
AA1287014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1287006 Supplier Thermo Scientific Chemicals Supplier No. 01287006
Only null left

CAS: 7759-02-6 | O4SSr | 183.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7759-02-6
Molecular Formula O4SSr
Molecular Weight (g/mol) 183.68
MDL Number MFCD00036274
InChI Key UBXAKNTVXQMEAG-UHFFFAOYSA-L
Synonym strontium sulfate, strontium sulphate, sulfuric acid, strontium salt 1:1, strontium 2+ ion sulfate, unii-7q3kx2l47f, strontiumsulfat, celestine, strontium 2+ sulfate, strontium sulfate 25g
PubChem CID 3084026
IUPAC Name strontium;sulfate
SMILES [O-]S(=O)(=O)[O-].[Sr+2]

Specifications

Melting Point 1600°C
Color White
Physical Form Powder
Quantity 5 g
Linear Formula SrSO4
Merck Index 14,8848
Formula Weight 183.68
Percent Purity 99.9965%
Grade Puratronic
Odor Odorless
Density 3.960 g/mL
Assay (metals basis)
Chemical Name or Material Strontium sulfate
Show More Show Less
EINECSNumber 231-850-2
RTECSNumber WT1210000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only